Order Entry
Puerto Rico
ContactUsLinkComponent
 
Diazepam 0.1 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77692-238
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Diazepam 0.1 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77692-238
Supplier:  LGC Standards
Supplier Number:  LGCAMP0023.00-02
New Product
CAS Number:  

Specifications

  • CAS No.:
    439-14-5
  • Description:
    Diazepam 0.1 mg/ml in Methanol
  • Size:
    1 ml
  • Formula:
    C₁₆H₁₃ClN₂O
  • Cat. No.:
    77692-238

Specifications

About this item