Verapamil Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal, LGC Standards
77693-647
New Product
:
- CAS No.:152-11-4
- Description:Verapamil Hydrochloride 1.0 mg/ml in Methanol (as free base)
- Size:1 ml
- Formula:C₂₇H₃₈N₂O₄
- Molecular Weight:491.07 g/mol
- Cat. No.:77693-647