Cimetidine 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77692-237
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:51481-61-9
- Description:Cimetidine 1.0 mg/ml in Methanol
- Size:1 ml
- Formula:C₁₀H₁₆N₆S
- Molecular Weight:252.34 g/mol
- Cat. No.:77692-237