Ambroxol Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal, LGC Standards
77692-235
New Product
:
- CAS No.:23828-92-4
- Description:Ambroxol Hydrochloride 1.0 mg/ml in Methanol (as free base)
- Size:1 ml
- Formula:C₁₃H₁₈Br₂N₂O.HCl
- Molecular Weight:414.57 g/mol
- Cat. No.:77692-235