Trichloroacetic acid-methyl ester 1000 µg/mL in Methyl-tert-butyl ether, Dr. Ehrenstorfer, LGC Standards
Catalog # 77693-513
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:598-99-2
- Description:Trichloroacetic acid-methyl ester 1000 µg/mL in Methyl-tert-butyl ether
- Size:1 ml
- Formula:C₃H₃Cl₃O₂
- Molecular Weight:177.41 g/mol
- Cat. No.:77693-513