Order Entry
Puerto Rico
ContactUsLinkComponent
 
4,4'-Dibromooctafluorobiphenyl 2000 μg/mL in Methyl-tert-butyl ether, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-925
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
4,4'-Dibromooctafluorobiphenyl 2000 μg/mL in Methyl-tert-butyl ether, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-925
Supplier:  LGC Standards
Supplier Number:  DRE-YA12240600MB
New Product
CAS Number:  

Specifications

  • CAS No.:
    10386-84-2
  • Description:
    4,4'-Dibromooctafluorobiphenyl 2000 μg/mL in Methyl-tert-butyl ether
  • Size:
    1 ml
  • Formula:
    C₁₂Br₂F₈
  • Molecular Weight:
    455.94 g/mol
  • Cat. No.:
    77691-925

Specifications

About this item