2,4,5-Trichlorophenol 100 µg/mL in Methanol, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-912
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:95-95-4
- Description:2,4,5-Trichlorophenol 100 µg/mL in Methanol
- Size:1 ml
- Formula:C₆H₃Cl₃O
- Molecular Weight:197.45 g/mol
- Cat. No.:77691-912