Order Entry
Puerto Rico
ContactUsLinkComponent
cis-Permethrin 100 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
cis-Permethrin 100 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
  77691-908
New Product
 :  
undefined
cis-Permethrin 100 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
  77691-908
 :  DRE-XA15990100CY
New Product
 :  

 

  • CAS No.:
    61949-76-6
  • Description:
    cis-Permethrin 100 µg/mL in Cyclohexane
  • Size:
    1 ml
  • Formula:
    C₂₁H₂₀Cl₂O₃
  • Cat. No.:
    77691-908