2,4-Dibromophenol 100 µg/mL in Methanol, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-888
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:615-58-7
- Description:2,4-Dibromophenol 100 µg/mL in Methanol
- Size:1 ml
- Formula:C₆H₄Br₂O
- Molecular Weight:251.91 g/mol
- Cat. No.:77691-888