Chlorthal-dimethyl 100 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
77691-874
New Product
:
- CAS No.:1861-32-1
- Description:Chlorthal-dimethyl 100 µg/mL in Cyclohexane
- Size:1 ml
- Formula:C₁₀H₆Cl₄O₄
- Molecular Weight:331.97 g/mol
- Cat. No.:77691-874