Atrazine 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-849
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:1912-24-9
- Description:Atrazine 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₈H₁₄ClN₅
- Molecular Weight:215.69 g/mol
- Cat. No.:77691-849