Abamectin 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-835
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:71751-41-2
- Description:Abamectin 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₄₈H₇₂O₁₄
- Molecular Weight:873.10 g/mol
- Cat. No.:77691-835
Specifications
About this item
Synonyms: (10E,14E,16E,22Z)-(1R,4S,5S',6S,6R',8R,12S,13S,20R,21R,24S)-6'[(S)-sec-butyl]-21,24-dihydroxy-5',11,13,22-tetramethyl-2-oxo-3,7,19-trioxatetracyclo[15.6.1.1 4.8.0 20.24]-pentacosa-10,14,16,22-tetraene-6-spiro-2'-(5',6'-dihydro-2'H-pyran)-12-yl 2,6-dideoxy-4,O-(2,6-dideoxy-3-O-methyl-a-L-arabinohexopyranosyl)-3-O-methyl-a-L-arabinohexopyranoside(i) mixture with (10E,14E,16E,22Z)-(1R,4S,5'S,6S,6'R,8R,12S,13S,20R,21R,24S)-21,22-dihydroxy-6'-isopropyl-5',11,13,22-tetramethyl-2-oxo-3,7,19-trioxatetracyclo[15.6.1.1 4.8.0 20.24]-pentacosa-10,14,16,22-tetraene-6-spiro-2'-(5',6'-dihydro-2'H-pyran)-12-yl 2,6-dideoxy-4-0-(2,6-dideoxy-3-O-methyl-a-L-arabinohexopyranosyl)-3-O-methyl-a-L-arabinohexopyranoside(ii) (4:1)