Order Entry
Puerto Rico
ContactUsLinkComponent
Chloroparaffin C14-C17 57% Cl 100 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
Chloroparaffin C14-C17 57% Cl 100 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
Catalog # 77679-224
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Chloroparaffin C14-C17 57% Cl 100 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
Catalog # 77679-224
Supplier:  LGC Standards
Supplier Number:  DRE-X23145700CY
New Product
CAS Number:  

Specifications

  • CAS No.:
    85535-85-9
  • Description:
    Chloroparaffin C14-C17 57% Cl 100 µg/mL in Cyclohexane
  • Size:
    10 ml
  • Cat. No.:
    77679-224

Specifications

About this item