Order Entry
Puerto Rico
ContactUsLinkComponent
Chloroparaffin C10-C13, 55,5% Cl 100 μg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
Chloroparaffin C10-C13, 55,5% Cl 100 μg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
Catalog # 77679-220
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Chloroparaffin C10-C13, 55,5% Cl 100 μg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
Catalog # 77679-220
Supplier:  LGC Standards
Supplier Number:  DRE-X23105500CY
New Product
CAS Number:  

Specifications

  • CAS No.:
    85535-84-8
  • Description:
    Chloroparaffin C10-C13, 55,5% Cl 100 μg/mL in Cyclohexane
  • Size:
    10 ml
  • Cat. No.:
    77679-220

Specifications

About this item