Order Entry
Puerto Rico
ContactUsLinkComponent
 
Benzo[a]pyrene D12 10 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
  77691-828
New Product
 :  
undefined
Benzo[a]pyrene D12 10 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
  77691-828
 :  DRE-LA20635100CY
New Product
 :  

 

  • CAS No.:
    63466-71-7
  • Description:
    Benzo[a]pyrene D12 10 µg/mL in Cyclohexane
  • Size:
    1 ml
  • Formula:
    C₂₀D₁₂
  • Cat. No.:
    77691-828