AHMI 10 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-817
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:15323-35-0
- Description:AHMI 10 µg/mL in Cyclohexane
- Size:1 ml
- Formula:C₁₇H₂₄O
- Molecular Weight:244.38 g/mol
- Cat. No.:77691-817