Order Entry
Puerto Rico
ContactUsLinkComponent
1-Methylfluoranthene 10 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
1-Methylfluoranthene 10 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
  77679-178
New Product
 :  
undefined
1-Methylfluoranthene 10 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
  77679-178
 :  DRE-L20892500CY
New Product
 :  

 

  • CAS No.:
    25889-60-5
  • Description:
    1-Methylfluoranthene 10 µg/mL in Cyclohexane
  • Size:
    10 ml
  • Formula:
    C₁₇H₁₂
  • Cat. No.:
    77679-178