5-Methylchrysene 10 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
Catalog # 77679-177
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:3697-24-3
- Description:5-Methylchrysene 10 µg/mL in Cyclohexane
- Size:10 ml
- Formula:C₁₉H₁₄
- Molecular Weight:242.32 g/mol
- Cat. No.:77679-177