2,4,6-Trichloroanisole 10 µg/mL in Isooctane, Dr. Ehrenstorfer, LGC Standards
Catalog # 77679-132
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:87-40-1
- Description:2,4,6-Trichloroanisole 10 µg/mL in Isooctane
- Size:10 ml
- Formula:C₇H₅Cl₃O
- Molecular Weight:211.47 g/mol
- Cat. No.:77679-132