Quintozene 10 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
Catalog # 77679-120
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:82-68-8
- Description:Quintozene 10 µg/mL in Cyclohexane
- Size:10 ml
- Formula:C₆Cl₅NO₂
- Molecular Weight:295.33 g/mol
- Cat. No.:77679-120