Propyzamide 10 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
Catalog # 77679-113
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:23950-58-5
- Description:Propyzamide 10 µg/mL in Cyclohexane
- Size:10 ml
- Formula:C₁₂H₁₁Cl₂NO
- Molecular Weight:256.13 g/mol
- Cat. No.:77679-113