Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Phorate 10 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
Phorate 10 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
Phorate 10 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
77679-109
:
LGC Standards
New Product
:
Phorate 10 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
77679-109
:
LGC Standards
:
DRE-L16080000CY
New Product
:
CAS No.:
298-02-2
Description:
Phorate 10 µg/mL in Cyclohexane
Size:
10 ml
Cat. No.:
77679-109