Pentachlorophenol 10 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
Catalog # 77679-105
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:87-86-5
- Description:Pentachlorophenol 10 µg/mL in Cyclohexane
- Size:10 ml
- Formula:C₆HCl₅O
- Molecular Weight:266.34 g/mol
- Cat. No.:77679-105