Order Entry
Puerto Rico
ContactUsLinkComponent
Mecoprop 10 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Mecoprop 10 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77679-089
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Mecoprop 10 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77679-089
Supplier:  LGC Standards
Supplier Number:  DRE-L14820000AL
New Product
CAS Number:  

Specifications

  • CAS No.:
    93-65-2
  • Description:
    Mecoprop 10 µg/mL in Acetonitrile
  • Size:
    10 ml
  • Formula:
    C₁₀H₁₁ClO₃
  • Molecular Weight:
    214.65 g/mol
  • Cat. No.:
    77679-089

Specifications

About this item