n-Heptyltin trichloride 10 µg/mL in Methyl-tert-butyl ether, Dr. Ehrenstorfer, LGC Standards
77679-080
New Product
:
- CAS No.:59344-47-7
- Description:n-Heptyltin trichloride 10 µg/mL in Methyl-tert-butyl ether
- Size:10 ml
- Formula:C₇H₁₅Cl₃Sn
- Cat. No.:77679-080