Ethylene thiourea 10 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
77679-072
New Product
:
- CAS No.:96-45-7
- Description:Ethylene thiourea 10 µg/mL in Acetonitrile
- Size:10 ml
- Formula:C₃H₆N₂S
- Molecular Weight:102.16 g/mol
- Cat. No.:77679-072