Difethialone 10 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77679-062
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:104653-34-1
- Description:Difethialone 10 µg/mL in Acetonitrile
- Size:10 ml
- Formula:C₃₁H₂₃BrO₂S
- Molecular Weight:539.4821 g/mol
- Cat. No.:77679-062