Dichlofenthion 10 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
Catalog # 77679-060
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:97-17-6
- Description:Dichlofenthion 10 µg/mL in Cyclohexane
- Size:10 ml
- Formula:C₁₀H₁₃Cl₂O₃PS
- Molecular Weight:315.16 g/mol
- Cat. No.:77679-060