Order Entry
Puerto Rico
ContactUsLinkComponent
gamma-Cyhalothrin 10 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
gamma-Cyhalothrin 10 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
Catalog # 77679-055
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
gamma-Cyhalothrin 10 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
Catalog # 77679-055
Supplier:  LGC Standards
Supplier Number:  DRE-L11859500CY
New Product
CAS Number:  

Specifications

  • CAS No.:
    76703-62-3
  • Description:
    gamma-Cyhalothrin 10 µg/mL in Cyclohexane
  • Size:
    10 ml
  • Formula:
    C₂₃H₁₉ClF₃NO₃
  • Cat. No.:
    77679-055

Specifications

About this item