Chlorfenapyr 10 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
Catalog # 77679-048
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:122453-73-0
- Description:Chlorfenapyr 10 µg/mL in Cyclohexane
- Size:10 ml
- Formula:C₁₅H₁₁BrClF₃N₂O
- Cat. No.:77679-048