Chloramphenicol 10 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77679-046
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:56-75-7
- Description:Chloramphenicol 10 µg/mL in Acetonitrile
- Size:10 ml
- Formula:Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
- Molecular Weight:323.13 g/mol
- Cat. No.:77679-046