Acetamiprid 10 µg/mL in Ethyl acetate, Dr. Ehrenstorfer, LGC Standards
Catalog # 77679-036
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:135410-20-7
- Description:Acetamiprid 10 µg/mL in Ethyl acetate
- Size:10 ml
- Formula:C₁₀H₁₁ClN₄
- Molecular Weight:222.67 g/mol
- Cat. No.:77679-036