Bromomethane (Methyl bromide) 100 µg/mL in Methanol, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-780
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:74-83-9
- Description:Bromomethane (Methyl bromide) 100 µg/mL in Methanol
- Size:1 ml
- Formula:BrCH₃
- Molecular Weight:94.94 g/mol
- Cat. No.:77691-780