Azadirachtin 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-777
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:11141-17-6
- Description:Azadirachtin 100 µg/mL in Acetonitrile
- Size:5x 1 ml
- Formula:C₃₅H₄₄O₁₆
- Molecular Weight:720.73 g/mol
- Cat. No.:77691-777