Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
4-Methoxy-1,3-phenylendiamine, Dr. Ehrenstorfer, LGC Standards
4-Methoxy-1,3-phenylendiamine, Dr. Ehrenstorfer, LGC Standards
77691-730
:
LGC Standards
New Product
:
4-Methoxy-1,3-phenylendiamine, Dr. Ehrenstorfer, LGC Standards
77691-730
:
LGC Standards
:
DRE-CA15081900
New Product
:
CAS No.:
615-05-4
Description:
4-Methoxy-1,3-phenylendiamine
Size:
100 mg
Formula:
C₇H₁₀N₂O
Molecular Weight:
138.17 g/mol
Cat. No.:
77691-730