Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
1,3-Dichloropropane, Dr. Ehrenstorfer, LGC Standards
1,3-Dichloropropane, Dr. Ehrenstorfer, LGC Standards
1,3-Dichloropropane, Dr. Ehrenstorfer, LGC Standards
77691-719
:
LGC Standards
New Product
:
1,3-Dichloropropane, Dr. Ehrenstorfer, LGC Standards
77691-719
:
LGC Standards
:
DRE-CA12481000
New Product
:
CAS No.:
142-28-9
Description:
1,3-Dichloropropane
Size:
250 mg
Formula:
ClH₂CCH₂CH₂Cl
Molecular Weight:
112.99 g/mol
Cat. No.:
77691-719