Acetic acid-tert-butyl ester, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-703
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:540-88-5
- Description:Acetic acid-tert-butyl ester
- Size:1 ml
- Formula:C₆H₁₂O₂
- Molecular Weight:116.16 g/mol
- Cat. No.:77691-703