DL-Valine, Dr. Ehrenstorfer, LGC Standards
Catalog # 77678-922
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:516-06-3
- Description:DL-Valine
- Size:100 mg
- Formula:C₅H₁₁NO₂
- Molecular Weight:117.15 g/mol
- Cat. No.:77678-922