$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Sulfanilamide, Dr. Ehrenstorfer, LGC Standards
Sulfanilamide, Dr. Ehrenstorfer, LGC Standards
77678-763
:
LGC Standards
New Product
:
Sulfanilamide, Dr. Ehrenstorfer, LGC Standards
77678-763
:
LGC Standards
:
DRE-C17000000
New Product
:
CAS No.:
63-74-1
Description:
Sulfanilamide
Size:
250 mg
Formula:
H₂NC₆H₄SO₂NH₂
Molecular Weight:
172.21 g/mol
Cat. No.:
77678-763