Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Sudan 4 D6 (naphthyl D6), Dr. Ehrenstorfer, LGC Standards
Sudan 4 D6 (naphthyl D6), Dr. Ehrenstorfer, LGC Standards
Sudan 4 D6 (naphthyl D6), Dr. Ehrenstorfer, LGC Standards
Catalog # 77678-743
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
Sudan 4 D6 (naphthyl D6), Dr. Ehrenstorfer, LGC Standards
Catalog # 77678-743
Supplier:
LGC Standards
Supplier Number:
DRE-C16986108
New Product
CAS Number:
Specifications
CAS No.:
1014689-18-9
Description:
Sudan 4 D6 (naphthyl D6)
Size:
10 mg
Cat. No.:
77678-743
Specifications
About this item
Product Details
Product Details