Scopolamine hydrobromide trihydrate, Dr. Ehrenstorfer, LGC Standards
Catalog # 77678-712
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:6533-68-2
- Description:Scopolamine hydrobromide trihydrate
- Size:100 mg
- Formula:C₁₇H₂₁NO₄
- Molecular Weight:384.27 g/mol
- Cat. No.:77678-712