Roxithromycin, Dr. Ehrenstorfer, LGC Standards
Catalog # 77678-702
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:80214-83-1
- Description:Roxithromycin
- Size:100 mg
- Formula:C₄₁H₇₆N₂O₁₅
- Molecular Weight:837.04 g/mol
- Cat. No.:77678-702