Rifampicin, Dr. Ehrenstorfer, LGC Standards
Catalog # 77678-697
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:13292-46-1
- Description:Rifampicin
- Size:100 mg
- Formula:C₄₃H₅₈N₄O₁₂
- Molecular Weight:822.95 g/mol
- Cat. No.:77678-697