Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Phthalic acid, bis-hexyl ester, Dr. Ehrenstorfer, LGC Standards
Phthalic acid, bis-hexyl ester, Dr. Ehrenstorfer, LGC Standards
Phthalic acid, bis-hexyl ester, Dr. Ehrenstorfer, LGC Standards
77678-578
:
LGC Standards
New Product
:
Phthalic acid, bis-hexyl ester, Dr. Ehrenstorfer, LGC Standards
77678-578
:
LGC Standards
:
DRE-C16173200
New Product
:
CAS No.:
84-75-3
Description:
Phthalic acid, bis-hexyl ester
Size:
100 mg
Formula:
C₂₀H₃₀O₄
Molecular Weight:
334.46 g/mol
Cat. No.:
77678-578