Patent Blue V, Dr. Ehrenstorfer, LGC Standards
Catalog # 77678-505
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:20262-76-4
- Description:Patent Blue V
- Size:250 mg
- Formula:C₂₇H₃₁N₂NaO₇S₂
- Molecular Weight:582.67 g/mol
- Cat. No.:77678-505