$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Parsol SLX (technical), Dr. Ehrenstorfer, LGC Standards
Parsol SLX (technical), Dr. Ehrenstorfer, LGC Standards
Catalog # 77678-504
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
Parsol SLX (technical), Dr. Ehrenstorfer, LGC Standards
Catalog # 77678-504
Supplier:
LGC Standards
Supplier Number:
DRE-C15894570
New Product
CAS Number:
Specifications
CAS No.:
207574-74-1
Description:
Parsol SLX (technical)
Size:
250 mg
Cat. No.:
77678-504
Specifications
About this item
Product Details
Product Details