Paracetamol, Dr. Ehrenstorfer, LGC Standards
Catalog # 77678-494
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:103-90-2
- Description:Paracetamol
- Size:250 mg
- Formula:C₈H₉NO₂
- Molecular Weight:151.16 g/mol
- Cat. No.:77678-494