$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
4-iso-Octylphenol-ethoxylate (technical), Dr. Ehrenstorfer, LGC Standards
4-iso-Octylphenol-ethoxylate (technical), Dr. Ehrenstorfer, LGC Standards
77678-473
:
LGC Standards
New Product
:
4-iso-Octylphenol-ethoxylate (technical), Dr. Ehrenstorfer, LGC Standards
77678-473
:
LGC Standards
:
DRE-C15712808
New Product
:
CAS No.:
9036-19-5
Description:
4-iso-Octylphenol-ethoxylate (technical)
Size:
250 mg
Cat. No.:
77678-473