Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Navy Blue 018112, Dr. Ehrenstorfer, LGC Standards
Navy Blue 018112, Dr. Ehrenstorfer, LGC Standards
Navy Blue 018112, Dr. Ehrenstorfer, LGC Standards
77678-414
:
LGC Standards
New Product
:
Navy Blue 018112, Dr. Ehrenstorfer, LGC Standards
77678-414
:
LGC Standards
:
DRE-C15492000
New Product
:
CAS No.:
118685-33-9
Description:
Navy Blue 018112
Size:
100 mg
Cat. No.:
77678-414