Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
tau-Fluvalinate, Dr. Ehrenstorfer, LGC Standards
tau-Fluvalinate, Dr. Ehrenstorfer, LGC Standards
tau-Fluvalinate, Dr. Ehrenstorfer, LGC Standards
Catalog # 77678-120
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
tau-Fluvalinate, Dr. Ehrenstorfer, LGC Standards
Catalog # 77678-120
Supplier:
LGC Standards
Supplier Number:
DRE-C13870000
New Product
CAS Number:
Specifications
CAS No.:
102851-06-9
Description:
tau-Fluvalinate
Size:
100 mg
Cat. No.:
77678-120
Specifications
About this item
Product Details
Product Details